Assay Details
| Gene Symbol: bsdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030463.1 | NP_001025634.1 | 10 - 11 | 1213 | 72 | |
| XM_012957089.1 | XP_012812543.1 | 10 - 11 | 1379 | 72 | ||
| XM_012957090.2 | XP_012812544.1 | 10 - 11 | 1323 | 72 | ||
| GenBank mRNA | BC091590.1 | - | 10 - 11 | 1213 | 72 | |
More Information
Set Membership: |
|

bsdc1
Genomic Map