Assay Details
| Gene Symbol: tmbim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030497.2 | NP_001025668.1 | 970 | 59 | ||
| GenBank mRNA | CX995578.1 | - | 9 - 10 | 531 | 59 | |
More Information
Set Membership: |
|

tmbim1
Genomic Map