Assay Details
| Gene Symbol: rab13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030508.1 | NP_001025679.1 | 4 - 5 | 475 | 101 | |
| GenBank mRNA | BC093468.1 | - | 4 - 5 | 475 | 101 | |
More Information
Set Membership: |
|

rab13
Genomic Map