Assay Details
| Gene Symbol: aldh1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032315.1 | NP_001027486.1 | 11 - 12 | 1389 | 54 | |
| GenBank mRNA | BC096010.1 | - | 11 - 12 | 1389 | 54 | |
More Information
Set Membership: |
|

aldh1a1
Genomic Map