Assay Details
| Gene Symbol: crata | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032318.1 | NP_001027489.1 | 12 - 13 | 1571 | 74 | |
| XM_012968519.2 | XP_012823973.1 | 12 - 13 | 1681 | 74 | ||
| GenBank mRNA | BC096389.1 | - | 12 - 13 | 1571 | 74 | |
More Information
Set Membership: |
|

crata
Genomic Map