Assay Details
| Gene Symbol: nmi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032331.1 | NP_001027502.2 | 12 - 13 | 1361 | 73 | |
| XM_012970498.2 | XP_012825952.1 | 12 - 13 | 1414 | 73 | ||
| XM_012970499.2 | XP_012825953.1 | 11 - 12 | 1344 | 73 | ||
| GenBank mRNA | BC098076.1 | - | 12 - 13 | 1361 | 73 | |
More Information
Set Membership: |
|

nmi
Genomic Map