Assay Details
| Gene Symbol: setd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032339.1 | NP_001027510.1 | 1 - 2 | 41 | 68 | |
| XM_012963846.2 | XP_012819300.1 | 1 - 2 | 186 | 68 | ||
| XM_018093866.1 | XP_017949355.1 | 1 - 2 | 187 | 68 | ||
| GenBank mRNA | BC098189.1 | - | 1 - 2 | 41 | 68 | |
More Information
Set Membership: |
|

setd7
Genomic Map