Assay Details
| Gene Symbol: trim8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032341.1 | NP_001027512.1 | 2 - 3 | 1194 | 81 | |
| XM_012966677.2 | XP_012822131.1 | 2 - 3 | 1349 | 81 | ||
| GenBank mRNA | BC098511.1 | - | 2 - 3 | 1194 | 81 | |
More Information
Set Membership: |
|

trim8
Genomic Map