Assay Details
| Gene Symbol: trim8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032341.1 | NP_001027512.1 | 6 - 7 | 1662 | 71 | |
| GenBank mRNA | BC098511.1 | - | 6 - 7 | 1662 | 71 | |
| Gene Symbol: LOC108645501 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018090689.1 | XP_017946178.1 | 2 - 3 | 217 | 71 | |
More Information
Set Membership: |
|

trim8
Genomic Map