Assay Details
| Gene Symbol: osgin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032344.1 | NP_001027515.1 | 4 - 5 | 483 | 66 | |
| XM_012964810.2 | XP_012820264.1 | 4 - 5 | 798 | 66 | ||
| XM_012964811.1 | XP_012820265.1 | 4 - 5 | 785 | 66 | ||
| XM_012964812.1 | XP_012820266.1 | 4 - 5 | 485 | 66 | ||
| GenBank mRNA | BC099667.1 | - | 4 - 5 | 483 | 66 | |
More Information
Set Membership: |
|

osgin2
Genomic Map