Assay Details
| Gene Symbol: vps51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032350.1 | NP_001027521.1 | 8 - 9 | 1948 | 61 | |
| GenBank mRNA | BC096636.1 | - | 8 - 9 | 1948 | 61 | |
More Information
Set Membership: |
|

vps51
Genomic Map