Assay Details
| Gene Symbol: tbx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032353.1 | NP_001027524.1 | 6 - 7 | 2189 | 102 | |
| GenBank mRNA | BC099621.1 | - | 6 - 7 | 2189 | 102 | |
More Information
Set Membership: |
|

tbx3
Genomic Map