Assay Details
| Gene Symbol: calu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213675.2 | NP_998840.1 | 5 - 6 | 699 | 71 | |
| XM_012958790.2 | XP_012814244.1 | 5 - 6 | 819 | 71 | ||
| GenBank mRNA | BC067980.2 | - | 5 - 6 | 699 | 71 | |
| CR942649.2 | - | 5 - 6 | 742 | 71 | ||
More Information
Set Membership: |
|

calu
Genomic Map