Assay Details
Gene Symbol: fam107b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007912.2 | NP_001007913.1 | 2 - 3 | 322 | 88 | |
XM_012958879.2 | XP_012814333.1 | 3 - 4 | 496 | 88 | ||
GenBank mRNA | BC080351.1 | - | 2 - 3 | 346 | 88 | |
CR848141.1 | - | 2 - 3 | 322 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
