Assay Details
| Gene Symbol: ppp1r14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007955.2 | NP_001007956.1 | 2 - 3 | 688 | 67 | |
| GenBank mRNA | BC080468.1 | - | 2 - 3 | 536 | 67 | |
| CR848110.2 | - | 2 - 3 | 688 | 67 | ||
More Information
Set Membership: |
|

ppp1r14a
Genomic Map