Assay Details
| Gene Symbol: ppp1r14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007955.2 | NP_001007956.1 | 3 - 4 | 733 | 76 | |
| GenBank mRNA | BC080468.1 | - | 3 - 4 | 581 | 76 | |
| CR848110.2 | - | 3 - 4 | 733 | 76 | ||
More Information
Set Membership: |
|

ppp1r14a
Genomic Map