Assay Details
| Gene Symbol: napa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011280.2 | NP_001011280.1 | 11 - 12 | 819 | 70 | |
| XM_012968301.1 | XP_012823755.1 | 11 - 12 | 1053 | 70 | ||
| GenBank mRNA | BC087994.1 | - | 11 - 12 | 848 | 70 | |
| CR926391.2 | - | 11 - 12 | 819 | 70 | ||
More Information
Set Membership: |
|

napa
Genomic Map