Assay Details
Gene Symbol: rbbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011394.2 | NP_001011394.2 | 2 - 3 | 226 | 83 | |
XM_018091215.1 | XP_017946704.1 | 2 - 3 | 339 | 83 | ||
GenBank mRNA | BC088588.1 | - | 2 - 3 | 212 | 83 | |
CR760337.2 | - | 2 - 3 | 226 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
