Assay Details
| Gene Symbol: rnps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015956.3 | NP_001015956.1 | 4 - 5 | 569 | 102 | |
| GenBank mRNA | BC118691.1 | - | 4 - 5 | 471 | 102 | |
| CR848461.2 | - | 4 - 5 | 537 | 102 | ||
| CX464844.2 | - | 4 - 5 | 568 | 102 | ||
More Information
Set Membership: |
|

rnps1
Genomic Map