Assay Details
Gene Symbol: c1orf52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015959.2 | NP_001015959.1 | 2 - 3 | 476 | 85 | |
GenBank mRNA | BC075604.1 | - | 2 - 3 | 496 | 85 | |
BC158908.1 | - | 2 - 3 | 513 | 85 | ||
BC167283.1 | - | 2 - 3 | 513 | 85 | ||
CR848445.2 | - | 2 - 3 | 476 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
