Assay Details
| Gene Symbol: lcmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016033.2 | NP_001016033.1 | 1 - 2 | 128 | 74 | |
| GenBank mRNA | BC157150.1 | - | 1 - 2 | 233 | 74 | |
| CR848122.2 | - | 1 - 2 | 128 | 74 | ||
More Information
Set Membership: |
|

lcmt1
Genomic Map