Assay Details
| Gene Symbol: ccdc43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016065.2 | NP_001016065.1 | 1 - 2 | 192 | 65 | |
| GenBank mRNA | BC118692.1 | - | 1 - 2 | 193 | 65 | |
| CR762269.2 | - | 1 - 2 | 192 | 65 | ||
More Information
Set Membership: |
|

ccdc43
Genomic Map