Assay Details
| Gene Symbol: elovl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016159.2 | NP_001016159.1 | 6 - 7 | 679 | 66 | |
| GenBank mRNA | BC135162.1 | - | 6 - 7 | 679 | 66 | |
| CR761931.2 | - | 6 - 7 | 679 | 66 | ||
More Information
Set Membership: |
|

elovl2
Genomic Map