Assay Details
| Gene Symbol: cdkn2aipnl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016247.2 | NP_001016247.1 | 2 - 3 | 333 | 99 | |
| GenBank mRNA | CR761768.2 | - | 2 - 3 | 333 | 99 | |
More Information
Set Membership: |
|

cdkn2aipnl
Genomic Map