Assay Details
| Gene Symbol: naif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016353.2 | NP_001016353.1 | 1 - 2 | 514 | 61 | |
| GenBank mRNA | BC128614.1 | - | 1 - 2 | 436 | 61 | |
| CR761320.2 | - | 1 - 2 | 514 | 61 | ||
More Information
Set Membership: |
|

naif1
Genomic Map