Assay Details
| Gene Symbol: ap4s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016360.2 | NP_001016360.1 | 2 - 3 | 270 | 94 | |
| XM_012968396.2 | XP_012823850.1 | 3 - 4 | 94 | |||
| XM_012968397.2 | XP_012823851.1 | 3 - 4 | 94 | |||
| GenBank mRNA | BC135702.1 | - | 3 - 4 | 310 | 94 | |
| CR761300.2 | - | 2 - 3 | 270 | 94 | ||
More Information
Set Membership: |
|

ap4s1
Genomic Map