Assay Details
| Gene Symbol: psma2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016382.2 | NP_001016382.1 | 1 - 2 | 54 | 87 | |
| GenBank mRNA | BC157163.1 | - | 1 - 2 | 168 | 87 | |
| CR761217.2 | - | 1 - 2 | 54 | 87 | ||
More Information
Set Membership: |
|

psma2
Genomic Map