Assay Details
| Gene Symbol: slc35e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016384.2 | NP_001016384.1 | 3 - 4 | 753 | 70 | |
| GenBank mRNA | BC170897.1 | - | 3 - 4 | 729 | 70 | |
| BC170901.1 | - | 3 - 4 | 729 | 70 | ||
| CR761211.2 | - | 3 - 4 | 753 | 70 | ||
More Information
Set Membership: |
|

slc35e1
Genomic Map