Assay Details
| Gene Symbol: commd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016404.2 | NP_001016404.1 | 4 - 5 | 560 | 85 | |
| GenBank mRNA | BC153686.2 | - | 4 - 5 | 524 | 85 | |
| CR761140.2 | - | 4 - 5 | 560 | 85 | ||
More Information
Set Membership: |
|

commd8
Genomic Map