Assay Details
| Gene Symbol: ube2s | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016438.2 | NP_001016438.1 | 3 - 4 | 394 | 67 | |
| GenBank mRNA | BC170651.1 | - | 3 - 4 | 384 | 67 | |
| BC170653.1 | - | 3 - 4 | 384 | 67 | ||
| CR761031.2 | - | 3 - 4 | 394 | 67 | ||
| CR761379.2 | - | 2 - 3 | 215 | 67 | ||
| CR762108.2 | - | 3 - 4 | 363 | 67 | ||
More Information
Set Membership: |
|

ube2s
Genomic Map