Assay Details
| Gene Symbol: wdr41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016529.2 | NP_001016529.1 | 4 - 5 | 493 | 60 | |
| XM_012962436.2 | XP_012817890.1 | 4 - 5 | 547 | 60 | ||
| GenBank mRNA | BC128996.1 | - | 4 - 5 | 474 | 60 | |
| CR926183.2 | - | 4 - 5 | 493 | 60 | ||
More Information
Set Membership: |
|

wdr41
Genomic Map