Assay Details
| Gene Symbol: mmgt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016553.2 | NP_001016553.1 | 2 - 3 | 146 | 94 | |
| GenBank mRNA | BC167281.1 | - | 2 - 3 | 201 | 94 | |
| BC170697.1 | - | 2 - 3 | 146 | 94 | ||
| BC170699.1 | - | 2 - 3 | 146 | 94 | ||
| CR760716.2 | - | 2 - 3 | 146 | 94 | ||
More Information
Set Membership: |
|

mmgt1
Genomic Map