Assay Details
| Gene Symbol: nudt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016613.2 | NP_001016613.1 | 9 - 10 | 595 | 56 | |
| XM_012959033.2 | XP_012814487.1 | 9 - 10 | 633 | 56 | ||
| XM_018092088.1 | XP_017947577.1 | 10 - 11 | 681 | 56 | ||
| GenBank mRNA | BC170594.1 | - | 8 - 9 | 585 | 56 | |
| BC170598.1 | - | 8 - 9 | 585 | 56 | ||
| CR760517.2 | - | 9 - 10 | 595 | 56 | ||
More Information
Set Membership: |
|

nudt5
Genomic Map