Assay Details
| Gene Symbol: eef1akmt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016660.2 | NP_001016660.1 | 2 - 3 | 326 | 76 | |
| GenBank mRNA | CR760304.2 | - | 2 - 3 | 326 | 76 | |
More Information
Set Membership: |
|

eef1akmt3
Genomic Map