Assay Details
| Gene Symbol: uqcrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016710.2 | NP_001016710.1 | 3 - 4 | 621 | 65 | |
| GenBank mRNA | CR760120.2 | - | 3 - 4 | 621 | 65 | |
More Information
Set Membership: |
|

uqcrc1
Genomic Map