Assay Details
| Gene Symbol: cdk2ap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016730.2 | NP_001016730.1 | 3 - 4 | 310 | 72 | |
| XM_018093477.1 | XP_017948966.1 | 3 - 4 | 493 | 72 | ||
| GenBank mRNA | BC084911.1 | - | 3 - 4 | 413 | 72 | |
| BC121523.1 | - | 3 - 4 | 329 | 72 | ||
| BC167332.1 | - | 3 - 4 | 521 | 72 | ||
| CR760030.2 | - | 3 - 4 | 310 | 72 | ||
More Information
Set Membership: |
|

cdk2ap1
Genomic Map