Assay Details
| Gene Symbol: desi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016799.2 | NP_001016799.1 | 1 - 2 | 97 | 58 | |
| GenBank mRNA | CR855615.2 | - | 1 - 2 | 97 | 58 | |
More Information
Set Membership: |
|

desi1
Genomic Map