Assay Details
| Gene Symbol: abca8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016876.2 | NP_001016876.1 | 8 - 9 | 908 | 76 | |
| GenBank mRNA | BC154873.1 | - | 9 - 10 | 960 | 76 | |
| BC169142.1 | - | 7 - 8 | 820 | 76 | ||
| CR848608.2 | - | 8 - 9 | 908 | 76 | ||
More Information
Set Membership: |
|

abca8
Genomic Map