Assay Details
| Gene Symbol: mri1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016877.2 | NP_001016877.1 | 5 - 6 | 998 | 77 | |
| GenBank mRNA | BC118767.1 | - | 5 - 6 | 956 | 77 | |
| CR848601.2 | - | 5 - 6 | 998 | 77 | ||
More Information
Set Membership: |
|

mri1
Genomic Map