Assay Details
| Gene Symbol: srsf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004913922.3 | XP_004913979.1 | 1 - 2 | 389 | 58 | |
| XM_004913924.3 | XP_004913981.1 | 1 - 2 | 389 | 58 | ||
| GenBank mRNA | CR848599.2 | - | 1 - 2 | 295 | 58 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004913924.3 transcripts.
Set Membership: |
|

srsf11
Genomic Map