Assay Details
| Gene Symbol: tmem150a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016894.2 | NP_001016894.1 | 6 - 7 | 521 | 74 | |
| XM_018092095.1 | XP_017947584.1 | 5 - 6 | 673 | 74 | ||
| GenBank mRNA | CR848545.2 | - | 6 - 7 | 521 | 74 | |
| CR942725.1 | - | 6 - 7 | 623 | 74 | ||
More Information
Set Membership: |
|

tmem150a
Genomic Map