Assay Details
| Gene Symbol: nsmce3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016930.2 | NP_001016930.1 | 5 - 6 | 396 | 68 | |
| XM_012954225.2 | XP_012809679.1 | 5 - 6 | 502 | 68 | ||
| XM_012954226.2 | XP_012809680.1 | 5 - 6 | 429 | 68 | ||
| GenBank mRNA | BC155975.1 | - | 5 - 6 | 401 | 68 | |
| CR762194.2 | - | 5 - 6 | 396 | 68 | ||
More Information
Set Membership: |
|

nsmce3
Genomic Map