Assay Details
| Gene Symbol: nsmce3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016930.2 | NP_001016930.1 | 9 - 10 | 674 | 98 | |
| XM_012954225.2 | XP_012809679.1 | 9 - 10 | 780 | 98 | ||
| XM_012954226.2 | XP_012809680.1 | 9 - 10 | 707 | 98 | ||
| GenBank mRNA | BC155975.1 | - | 9 - 10 | 679 | 98 | |
| CR762194.2 | - | 9 - 10 | 674 | 98 | ||
More Information
Set Membership: |
|

nsmce3
Genomic Map