Assay Details
| Gene Symbol: cmss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016943.2 | NP_001016943.1 | 4 - 5 | 381 | 81 | |
| GenBank mRNA | BC135225.1 | - | 4 - 5 | 435 | 81 | |
| CR761873.2 | - | 4 - 5 | 381 | 81 | ||
More Information
Set Membership: |
|

cmss1
Genomic Map