Assay Details
| Gene Symbol: tdp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016944.2 | NP_001016944.1 | 1 - 2 | 212 | 69 | |
| XM_012964761.1 | XP_012820215.1 | 1 - 2 | 222 | 69 | ||
| GenBank mRNA | BC166146.1 | - | 1 - 2 | 214 | 69 | |
| CR761829.2 | - | 1 - 2 | 212 | 69 | ||
More Information
Set Membership: |
|

tdp2
Genomic Map