Assay Details
| Gene Symbol: cox4i2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016945.2 | NP_001016945.1 | 115 | 76 | ||
| GenBank mRNA | BC155001.1 | - | 152 | 76 | ||
| CR761813.2 | - | 115 | 76 | |||
More Information
Set Membership: |
|

cox4i2
Genomic Map