Assay Details
| Gene Symbol: eif2b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016997.3 | NP_001016997.2 | 5 - 6 | 682 | 86 | |
| GenBank mRNA | BC157160.1 | - | 5 - 6 | 637 | 86 | |
| CR848137.2 | - | 5 - 6 | 639 | 86 | ||
More Information
Set Membership: |
|

eif2b3
Genomic Map