Assay Details
| Gene Symbol: hras | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017003.2 | NP_001017003.1 | 5 - 6 | 537 | 75 | |
| XM_012961161.2 | XP_012816615.1 | 6 - 7 | 767 | 75 | ||
| XM_012961162.2 | XP_012816616.1 | 3 - 4 | 383 | 75 | ||
| XM_018093060.1 | XP_017948549.1 | 2 - 3 | 300 | 75 | ||
| XM_018093061.1 | XP_017948550.1 | 3 - 4 | 321 | 75 | ||
| GenBank mRNA | CR762314.2 | - | 5 - 6 | 537 | 75 | |
More Information
Set Membership: |
|

hras
Genomic Map