Assay Details
| Gene Symbol: march2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017006.2 | NP_001017006.1 | 2 - 3 | 295 | 95 | |
| XM_018093634.1 | XP_017949123.1 | 2 - 3 | 256 | 95 | ||
| GenBank mRNA | BC153690.1 | - | 2 - 3 | 403 | 95 | |
| CR762288.2 | - | 2 - 3 | 295 | 95 | ||
More Information
Set Membership: |
|

march2
Genomic Map