Assay Details
| Gene Symbol: ndufa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017047.2 | NP_001017047.1 | 2 - 3 | 93 | 79 | |
| GenBank mRNA | CR761851.2 | - | 2 - 3 | 93 | 79 | |
More Information
Set Membership: |
|

ndufa3
Genomic Map